Learn More
Thermo Scientific™ HisPur™ Ni-NTA Spin Columns, 3 mL
Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.
Brand: Thermo Scientific™ 88226
168.27 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.
The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.
Highlights:
High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
Versatile – purify proteins using native or denaturing conditions
Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
Cost-effective – reuse the same batch of resin at least five times
Easy to use – pre-formulated buffers available for kit formats
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin
Includes:
Kits contain HisPur Ni-NTA Columns
Specifications
3 mL | |
Store at 4°C | |
HisPur™ | |
His-Tagged Protein | |
Divalent Nickel, Metal Chelate | |
Affinity, Agarose Resin |
HisPur Ni-NTA Spin Columns, 3mL | |
Liquid Suspension | |
5 Columns | |
Spin Column | |
Spin Column |
For Research Use Only. Not for use in diagnostic procedures.