missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit, 3 mL

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

Brand:  Thermo Scientific™ 88229

Code : PN

Additional Details : Weight : 0.43000kg Transport : UN number : 3267 Chem class : 8 Pack group : III

Product Code. 11892104

  • £379.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin

  • Versatile – purify proteins using native or denaturing conditions

  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives

  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges

  • Cost-effective – reuse the same batch of resin at least five times

  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns

Specifications

Specifications

1 mL/Column
Sufficient For: Purifying up to 180 mg of His-tagged protein per column
• HisPur Ni-NTA Spin Columns, 3 mL, 5 columns
• Phosphate-buffered Saline (10X), 30 mL
• Imidazole (2M), 25 mL
• Column Plugs, 5 plugs

Store at 4°C.
5 Column Kit
Spin Column Kit
180 mg of His-Tagged Protein per Column
Affinity, Agarose Resin
HisPur Ni-NTA Spin Purification Kit, 3mL
HisPur™
His-Tagged Protein
Divalent Nickel, Metal Chelate
Spin Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.