missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Resin

Product Code. 10459164 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Unit Size:
100mL
10mL
500mL
This item is not returnable. View return policy

Product Code. 10459164

Brand: Thermo Scientific™ 88223

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
  • Versatile – purify proteins using native or denaturing conditions
  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
  • Cost-effective – reuse the same batch of resin at least five times
  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns (store at 4°C)

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) Up to 60 mg of 6xHis-Tagged Protein Binding per mL Resin
Description HisPur Ni-NTA Resin
Content And Storage Store at 4°C
Form Liquid Suspension
Product Line HisPur
Quantity 500 mL
Target His-Tagged Protein
Type Resin
Stationary Phase Divalent Nickel, Metal Chelate
Format Bottle
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.