Learn More
Thermo Scientific™ HisPur™ Ni-NTA Superflow Agarose
Shop All Thermo Scientific ProductsDescription
Perform FPLC purification of poly-histidine-tagged proteins with Thermo Scientific™ HisPur Ni-NTA Superflow Agarose, a nitrilotriacetic acid (NTA) modified Superflow 6 support charged with divalent nickel (Ni+2).
Additional Information and Online Ordering
Highlights:
High capacity – binds greater than 20mg of His-tagged GFP per mL of resin at a linear flow rate of 300cm/hour
High purity – provides greater than 80% purity of eluted fractions when used to purify from lysates
Versatile – can be used to purify proteins under native and denaturing conditions
Robust – highly crosslinked beads tolerate linear flow rates up to 1200cm/hour
Compatible – maintains function in a wide variety of chemicals and pH conditions
Cost effective – competitively priced resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of polyhistidine tagged proteins
Specifications
Specifications
| Capacity (Metric) | 20 mg His-Tagged GFP Binding per mL Resin |
| Content And Storage | Store at 4°C |
| Form | Liquid Suspension |
| Product Line | HisPur |
| Quantity | 50 mL |
| Target | His-Tagged Protein |
| Type | Agarose |
| Stationary Phase | Divalent Nickel, Metal Chelate |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
Safety and Handling
- HisPur Ni-NTA Superflow Agarose, 50 mL.
- Danger
- Flammable liquid Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Carcinogenicity Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- MIXTURE LIST-Contains : Ethyl alcohol, Nickel(II) sulfate hexahydrate(1:1:6)
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.