missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HistoStar™ Heated Forceps
Epredia™ Heated Forceps work in conjunction with the Epredia™ HistoStar™ Embedding Workstation. Multiple sizes are available.
Brand: Epredia™ A81010033
601.60 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Epredia™ HistoStar™ Embedding Workstation | |
Heated Forceps | |
Red; 1 mm tip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction