Learn More
Epredia™ HistoStar™ Heated Forceps
Epredia™ Heated Forceps work in conjunction with the Epredia™ HistoStar™ Embedding Workstation. Multiple sizes are available.
Brand: Epredia™ A81010035
Code : 50
Additional Details : Weight : 0.50000kg
Specifications
Heated Forceps | |
Epredia™ HistoStar™ Embedding Workstation | |
Blue; 4 mm tip |