Learn More
Cytiva HisTrap™ excel Prepacked Columns
For capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants. Cytiva Lifescience™ HisTrap™ excel Prepacked Columns are designed for fast and convenient purifications.
Brand:  Cytiva 17-3712-06
 View more versions of this product
Product Code. 12683466
  • £748.25 / Pack of 5
Edge
Explore more special offers
Description
Show More
Specifications
Specifications
5 mL
16 mm
5 to 20 mL/min.
2 to 12
4°C to 30°C
5 x 5 mL
0.6 in.
HisTrap excel columns are prepacked with Ni Sepharose excel affinity resins for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).
Polypropylene
6% Highly Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
Pre-packed Column
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva HisTrap™ excel Prepacked Columns > Flow rate: less-than 20mL/min; Bed dimensions: 16 x 25mm; Packsize: 5 x 5mL