missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HisTrap™ FF Columns: 5 x 5mL

Optimize purifications of histidine-tagged proteins directly from homogenized, unclarified cells HisTrap™ FF Columns: 5 x 5mL are prepacked with precharged Ni Sepharose™ 6 Fast Flow media.

Brand:  Cytiva 17-5255-01


Product Code. 10571680

  • £552.00 / 25mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
  • Column material: polypropylene
  • Bed dimensions: 7 × 25mm
  • Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
  • Compatible with wide range of reducing agents, detergents, denaturants, and other additives
  • Simple operation with syringe, perstaltic pump, or chromatographic system
  • Columns may be connected in series to increase capacity
Medium
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
7 mm
<4 mL/min.
3 to 12
4°C to 30°C, 20% Ethanol
5 x 5 mL
0.2 in.
HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).
Polypropylene
6% Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
Pre-packed Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, Nickel
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers