missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HisTrap™ FF Columns: 7 x 25mm
Optimized for purification of histidine-tagged proteins directly from homogenized, unclarified cells
Brand: Cytiva 17-5255-02
Code : GX
Additional Details : Weight : 1.52000kg
Description
- Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
- Column material: polypropylene
- Bed dimensions: 7 × 25mm
- Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
- Compatible with wide range of reducing agents, detergents, denaturants, and other additives
- Simple operation with syringe, perstaltic pump, or chromatographic system
- Columns may be connected in series to increase capacity
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
Specifications
5 mL | |
7 mm | |
Polypropylene | |
6% Cross-linked Agarose | |
5 bar | |
Pre-packed Column |
0.2 in. | |
<4 mL/min. | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
100 x 5 mL |