missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HisTrap™ FF Crude Prepacked Columns
Purify histidine-tagged proteins directly from homogenized, unclarified cell lysates. Cytiva Life Sciences™ HisTrap™ FF Crude Prepacked Columns offer the convenience of simple, one-step purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
£176.00 - £7181.50
Specifications
Material | Polypropylene |
---|---|
pH Range | 3 to 12 |
Support Type | 6% Cross-linked Agarose |
Content And Storage | 4°C to 30°C |
Particle Size | 45 to 165 μm |
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
11723219
|
Cytiva
17-5286-01 |
5 x 5 mL | <20 mL/min. |
£599.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10685355
|
Cytiva
11-0004-58 |
60 mL | <4 mL/min. |
£176.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
11515674
|
Cytiva
11-0004-59 |
100 x 1 mL | <4 mL/min. | N/A | |||||
11800083
|
Cytiva
17-5286-02 |
100 x 5 mL | <20 mL/min. | N/A | |||||
Description
- Prepacked with Ni Sepharose™ 6 Fast Flow media
- Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
- 1 and 5mL columns
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Polypropylene | |
6% Cross-linked Agarose | |
45 to 165 μm |
3 to 12 | |
4°C to 30°C | |
Pre-packed Column |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains Nickel. May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol