Learn More
Cytiva HiTrap™ ANX Sepharose Fast Flow IEX Columnns
Weak anion exchanger for small scale purification of higher molecular weight protein
Brand:  Cytiva 17-5163-01
 View more versions of this product
Product Code. 11575025
  • £299.00 / Pack of 5
Explore more special offers
Description
Description
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap ANX FF (High Sub)
<20 mL/min.
4°C to 30°C in 20% Ethanol
5 mL
0.6 in.
HiTrap ANX (high sub) FF is prepacked with ANX (high sub) Sepharose Fast Flow, and is a weak anion exchanger for small scale purification of higher molecular weight protein as well as screening of binding and elutions conditions.
16 mm
5 x 5 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.