missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ ANX Sepharose Fast Flow IEX Columnns

Weak anion exchanger for small scale purification of higher molecular weight protein

£136.00 - £299.00

Specifications

Description HiTrap ANX FF (High Sub)
For Use With (Application) HiTrap ANX (high sub) FF is prepacked with ANX (high sub) Sepharose Fast Flow, and is a weak anion exchanger for small scale purification of higher molecular weight protein as well as screening of binding and elutions conditions.
Content And Storage 4°C to 30°C in 20% Ethanol
View More Specs

Products 2
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
10450895
View Documents Promotion Details
Cytiva
17-5162-01
1 mL 1 x 5 mL
£136.00
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
11575025
View Documents
Cytiva
17-5163-01
5 mL 5 x 5 mL
£299.00
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential

Specifications

Specifications

HiTrap ANX FF (High Sub)
HiTrap ANX (high sub) FF is prepacked with ANX (high sub) Sepharose Fast Flow, and is a weak anion exchanger for small scale purification of higher molecular weight protein as well as screening of binding and elutions conditions.
4°C to 30°C in 20% Ethanol
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.