missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Blue HP Columns

For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins

Brand:  Cytiva 17-0413-01


 View more versions of this product

Product Code. 11758448

  • £192.98 / Each
Estimated Shipment: 05-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Blue Sepharose High Performance media
  • Cibacron Blue F3G-A, ligand, is covalently attached to the 6% agarose medium
  • Stable over pH range 4 to 12 for use and 3 to 13 for cleaning

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 x 5mL
34 μm
5mL
1.6cm
0.98 in.
4mg Cibacron Blue F3G-A/mL medium
42psi
6% Highly Cross-linked Agarose
4° to 30°C
<20 mL/min.
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Blue HP Columns
Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins
2.5cm
Polypropylene
4 to 12
Albumin, interferon, a broad range of nucleotide-dependent Enzymes, a2-macroglobulin, coagulation facto rs and related Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hitrap blue HP 5ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers