Learn More
Cytiva HiTrap™ Butyl FF Columns
Prepacked columns for convenient and fast separations
Brand: Cytiva 17-5197-01
Code : GE
Additional Details : Weight : 2.14100kg
Description
- Flexible design for use with a syringe, laboratory pump or chromatography system
- Available in 1mL and 5mL sizes
- High flowrates and high capacities
- Easy scale-up achieved by connecting columns in series
- Maximum flowrates: 4mL/min. for 1mL columns and 20mL/min. for 5mL columns
Specifications
5 mL | |
16 mm | |
< 5 mL/min | |
5 x 5 mL | |
25 mm | |
Polypropylene | |
5 bar [0.5 MPa] (70 psi) | |
3 to 13 | |
4°C to 30°C, 20% Ethanol |
0.629 in. | |
HiTrap Butyl FF | |
For Screening of Selectivity, Binding and Elution Conditions, as Well as Small Scale Purifications | |
0.984 in. | |
Butyl | |
Butyl Sepharose 4 Fast Flow | |
45 to 165 μm | |
4% Cross-linked Agarose |
Safety and Handling
- HiTrap Butyl FF 5 ml, 5 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.