missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Butyl-S FF Columns

Designed for purification of relatively strong hydrophilic molecules at low salt concentrations

Brand:  Cytiva 17-0978-13


 View more versions of this product

Product Code. 10481085

  • £195.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Useful for removal of strong hydrophobic contaminants, such as lipoproteins, lipids, pigments and more from biological samples
  • Used successfully for large-scale purification of recombinant Hepatitis B virus surface antigen (rec-HBsAg) from CHO cells
  • Robust production with high reproducibility

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
7 mm
< 1 mL/min
5 x 1 mL
Polypropylene
45 to 165 μm
6% Cross-linked Agarose
0.275 in.
HiTrap Butyl-S FF column
For Screening of Selectivity, Binding and Elution Conditions, as Well as Small Scale Purifications
Butyl-S
5 bar [0.5 MPa] (70 psi)
3 to 13
4°C to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Butyl-S FF 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.