missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Butyl-S FF Columns
Designed for purification of relatively strong hydrophilic molecules at low salt concentrations
£189.00 - £256.11
Specifications
Product Type | HiTrap Butyl-S FF column |
---|---|
Ligand Type | Butyl-S |
Material | Polypropylene |
Particle Size | 45 to 165 μm |
pH Range | 3 to 13 |
Product Code | Brand | Diameter (English) Inner | Quantity | Flow Rate | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Quantity | Flow Rate | Price | Quantity & Availability | |||
10481085
|
Cytiva
17-0978-13 |
0.275 in. | 5 x 1 mL | < 1 mL/min |
£189.00
Pack of 5 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||
11584955
|
Cytiva
17-0978-14 |
0.629 in. | 5 x 5 mL | 5 mL/min |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Useful for removal of strong hydrophobic contaminants, such as lipoproteins, lipids, pigments and more from biological samples
- Used successfully for large-scale purification of recombinant Hepatitis B virus surface antigen (rec-HBsAg) from CHO cells
- Robust production with high reproducibility
Specifications
HiTrap Butyl-S FF column | |
Polypropylene | |
3 to 13 |
Butyl-S | |
45 to 165 μm | |
6% Cross-linked Agarose |