missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Adhere Columns

Designed to remove contaminants in post-protein A purification

Brand:  Cytiva 28-4058-46

745.75 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 11505135

  • £785.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Capto adhere media, a strong anion exchanger with multimodal functionality
  • High capacity and productivity in flow-through mode: 0.09 to 0.12mmol Cl-/mL medium ionic capacity
  • Wide operational window of pH and conductivity: working pH range 3 to 12
  • Two-step process reduces time spent and operating costs

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffer
5mL
16mm
0.98 in.
<20 mL/min.
N-benzyl-n-methyl Ethanolamine
3 to 12
Highly Cross-linked Agarose
Affinity Column
HiTrap Capto adhere column
0.62 in.
5 x 5mL
25mm
Polypropylene
44 bar
75μm
4° to 30°C
Easy screening and convenient process development
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Capto adhere 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.