missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto MMC Columns

HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.

Brand:  Cytiva 11-0032-75


 View more versions of this product

Product Code. 11748408

  • £549.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High dynamic binding capacity at high conductivity.
  • Selectivity allows binding in the presence of salt.
  • High volume throughput.
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffer
5mL
16mm
0.98 in.
300kPa at 600cm/hr.
3 bar
75μm
4° to 30°C
Bind Proteins at the conductivity of the feed material and to solve specific purification challenges
HiTrap Capto MMC column
0.62 in.
5 x 5mL
25mm
Polypropylene
2 to 12
Highly Cross-linked Agarose
Affinity Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hitrap capto MMC 5ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers