missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ Q IEX Columns
Strong quaternary ammonium (Q) anion exchanger
Brand: Cytiva 11-0013-02
Code : GE
Additional Details : Weight : 0.10000kg
Description
- High-throughput medium for capture and intermediate purification
- For process-scale purification
- Combined properties of high binding capacity, high flowrate and low back-pressure properties reduce process cycle times and improve productivity
- Rigid agarose matrix enables high bed heights and purification of viscous samples at high flowrates
- High dynamic binding capacity is achieved through dextran surface extender coating on the agarose matrix
Specifications
HiTrap Capto Q | |
0.2 in. | |
< 4 mL/min. | |
Quaternary amine | |
7 mm | |
90 μm (d50V) | |
5 x 1 mL |
Polypropylene | |
Highly cross-linked agarose with dextran surface extender | |
HiTrap Capto Q chromatography column is prepacked with a strong anion exchange modern resin for small scale protein purification. | |
4°C to 30°C in 20% Ethanol | |
1 mL | |
2 to 12 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Capto Q 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.