missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ S IEX Columns

Strong sulfoethyl(S) cation exchanger

Brand:  Cytiva 17-5441-22

Code : GE

Additional Details : Weight : 0.50000kg


 View more versions of this product

Product Code. 10034484

  • £150.00 / Pack of 5
Estimated Shipment: 06-08-2024
to see stock.

Description

Description

  • High-throughput medium for capture and intermediate purification
  • Useful for process-scale purification
  • Combined properties of high binding capacity, high flowrate and low back-pressure properties reduce process cycle times and improve productivity
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flowrates
  • High dynamic binding capacity is achieved through dextran surface extender coating on the agarose matrix

Specifications

Specifications

HiTrap Capto S, 5 x 1 mL
0.2 in.
< 4 mL/min.
Sulfonate group
7 mm
90 μm (d50V)
5 x 1 mL
Polypropylene
Highly cross-linked agarose with dextran surface extender
HiTrap Capto S chromatography column is prepacked with a strong cation exchange modern resin for small scale protein purification.
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
1 mL
4 to 12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Capto S 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers