missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap Capto S ImpAct Prepacked Columns
HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).
£191.00 - £729.00
Specifications
Product Type | HiTrap Capto S ImpAct Prepacked Column |
---|---|
Material | High-flow agarose |
pH Range | 4 to 12 |
Particle Size | 50μm |
Column Type | Ion Exchange Column |
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
15361642
|
Cytiva
17371751 |
5 x 1mL | 4mL/min |
£191.00
Pack of 5 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15371642
|
Cytiva
17371755 |
5 x 5mL | 20mL/min |
£729.00
Pack of 5 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||||
Description
- Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
- High binding capacity, > 100 mg MAb/mL medium.
- Efficient aggregate removal at high load of monoclonal antibodies.
- High-resolution intermediate purification and polishing based on the well-established Capto platform.
- Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
Specifications
HiTrap Capto S ImpAct Prepacked Column | |
4 to 12 | |
Ion Exchange Column |
High-flow agarose | |
50μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Capto S ImpAct, 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.