missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HiTrap™ Chromatography Columns with MabSelect™ VL Protein L Resin

Protein L affinity chromatography columns for capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

Brand:  Cytiva™ 17542054

Additional Details : CAS Number : 64-17-5 Weight : 0.13300kg

 View more versions of this product

Product Code. 17817774

  • £3250.00 / 25mL
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • Redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability
  • High dynamic binding capacity for efficient capture of target molecules
  • Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
  • Provides good resolution for product-related impurities in the capture of bispecific antibodies

As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.

Specifications

Specifications

5 mL
16 mm
1 in.
<20 mL/min.
Affinity Column
0.63 in.
5 x 5 mL
25 mm
5 bar
Antibody Affinity Chromatography
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers