missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Con A 4B Columns
For separation and purification of glycoproteins, polysaccharides and glycolipids
Brand: Cytiva 28952085
View more versions of this product
This item is not returnable.
View return policy
Description
- Prepacked with Con A Sepharose™ 4B media
- Bed volume: 1mL and 5mL
- Bed dimensions: 7 x 25mm (1mL), 16 x 25mm (5mL)
- Column material: Polypropylene (PP)
- Max flow rate: 4mL/min (1mL), 20mL/min (5mL)
- Ligand: Concanavalin A (Con A)
- Matrix: 4% agarose
- Average particle size: 90μm
- pH stability working range: 4 to 9
Specifications
Aqueous Buffer | |
1mL | |
7mm | |
0.98 in. | |
<4 mL/min. | |
Concanavalin A (13mg Con A/mL drained medium) | |
4 to 9 | |
4% Agarose | |
Ion Exchange Column |
HiTrap Con A Sepharose 4B column | |
0.27 in. | |
5 x 1mL | |
25mm | |
Polypropylene | |
3 bar | |
45 to 165μm | |
4° to 8°C | |
Convenient separation and purification of Glycoproteins, Polysaccharides and Glycolipids |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Con A 4B 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction