missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Con A 4B Columns

For separation and purification of glycoproteins, polysaccharides and glycolipids

Brand:  Cytiva 28-9520-96


 View more versions of this product

Product Code. 11731417

  • £641.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Con A Sepharose™ 4B media
  • Bed volume: 1mL and 5mL
  • Bed dimensions: 7 x 25mm (1mL), 16 x 25mm (5mL)
  • Column material: Polypropylene (PP)
  • Max flow rate: 4mL/min (1mL), 20mL/min (5mL)
Media
  • Ligand: Concanavalin A (Con A)
  • Matrix: 4% agarose
  • Average particle size: 90μm
  • pH stability working range: 4 to 9

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffer
5mL
16mm
0.98 in.
<20 mL/min.
Concanavalin A (13mg Con A/mL drained medium)
4 to 9
4% Agarose
Ion Exchange Column
HiTrap Con A Sepharose 4B column
0.62 in.
5 x 5mL
25mm
Polypropylene
3 bar
45 to 165μm
4° to 8°C
Convenient separation and purification of Glycoproteins, Polysaccharides and Glycolipids
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Con A 4B

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.