missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ DEAE Sepharose Fast Flow IEX Columns

Weak anion exchanger for small-scale protein purifications.  Cytiva Lifescience™ HiTrap™ DEAE Sepharose Fast Flow IEX Columns are convenient and rapid.

Brand:  Cytiva 17-5055-01

Code : GE

Additional Details : Weight : 0.09800kg


 View more versions of this product

Product Code. 10044394

  • £132.00 / Pack of 5
Estimated Shipment: 02-08-2024
to see stock.

Description

Description

  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential

Specifications

Specifications

HiTrap DEAE FF
< 4 mL/min.
4°C to 30°C in 20% Ethanol
1 mL
0.2 in.
For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules
7 mm
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap DEAE FF 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers