missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Desalting Prepacked Columns

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

Brand:  Cytiva 29048684


 View more versions of this product

Product Code. 15147956

This item is not returnable. View return policy

Description

Description

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20μm
  • pH stability working range: 2 to 13

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
>20 μm (dry), 15 to 88 μm (wet)
For rapid convenient desalting and buffer exchange
4°C to 30°C in 20% Ethanol
Reusable
HiTrap Desalting, 1 x 5 mL
0.62 in.
5 mL
16 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Desalting 5 ml, 1 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers