missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Heparin HP Columns

Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.

Brand:  Cytiva 17-0407-03


 View more versions of this product

Product Code. 11738448

  • £935.00 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stable over pH range 5 to 10
  • Tolerates commonly used aqueous buffers
  • Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 5mL
34 μm
5mL
1.6cm
0.98 in.
10mg Heparin/mL medium
42psi
6% Highly Cross-linked Agarose
4° to 30°C
<20 mL/min.
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Heparin HP Column
Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
2.5cm
Polypropylene
5 to 10
DNA-binding Proteins, Coagulation Factors, Lipoproteins, Protein Synthesis Factors
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Heparin HP 5 ml, 1 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers