missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ IMAC Columns

For optimizing purification of histidine-tagged proteins when Ni +2 is not the best choice of metal ion

Brand:  Cytiva 17-0921-04

Code : GE

Additional Details : Weight : 0.14200kg


 View more versions of this product

Product Code. 11758498

  • £522.00 / Pack of 5
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Conveniently charge with your metal of choice
  • Can be used with a syringe, peristaltic pump, or liquid chromatography system
  • Available prepacked with IMAC Sepharose™ High Performance (HP), with small bead size for high performance purification, or IMAC Sepharose 6 Fast Flow (FF), with larger bead size for easy scaleup
  • High binding capacity: ≥40mg histidine-taged protein/mL medium (protein/metal ion dependent)
  • Stable in the presence of all commonly used buffers
  • Available in 1mL and 5mL sizes

Specifications

Specifications

5 mL
<4 mL/min.
Prepacked with IMAC Sepharose 6 Fast flow media
Polypropylene
3 to 12
HiTrap IMAC Fast Flow Column
5 x 5 mL
0.62 in.
16 mm
Syringe, peristaltic pump or liquid chromato graphy system
42 psi
90 μm
4°C to 30°C
For optimizing purification of His-tagged Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap IMAC FF, 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers