missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ IMAC Columns
For optimizing purification of histidine-tagged proteins when Ni +2 is not the best choice of metal ion
£152.00 - £653.00
Specifications
Flow Rate | <4 mL/min. |
---|---|
For Use With (Equipment) | Syringe, peristaltic pump or liquid chromato graphy system |
Material | Polypropylene |
Max. Pressure | 42 psi |
pH Range | 3 to 12 |
Includes: Instructions, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems
Product Code | Brand | Diameter (English) Inner | Quantity | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Quantity | Particle Size | Price | Quantity & Availability | |||
10535735
|
Cytiva
17-0920-03 |
0.27 in. | 5 x 1 mL | 34 μm |
£193.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11514955
|
Cytiva
17-0920-05 |
0.62 in. | 5 x 5 mL | 34 μm |
£653.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10697275
|
Cytiva
17-0921-02 |
0.27 in. | 5 x 1 mL | 90 μm |
£152.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11758498
|
Cytiva
17-0921-04 |
0.62 in. | 5 x 5 mL | 90 μm |
£522.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
Description
- Conveniently charge with your metal of choice
- Can be used with a syringe, peristaltic pump, or liquid chromatography system
- Available prepacked with IMAC Sepharose™ High Performance (HP), with small bead size for high performance purification, or IMAC Sepharose 6 Fast Flow (FF), with larger bead size for easy scaleup
- High binding capacity: ≥40mg histidine-taged protein/mL medium (protein/metal ion dependent)
- Stable in the presence of all commonly used buffers
- Available in 1mL and 5mL sizes
Specifications
<4 mL/min. | |
Polypropylene | |
3 to 12 | |
For optimizing purification of His-tagged Proteins |
Syringe, peristaltic pump or liquid chromato graphy system | |
42 psi | |
4°C to 30°C |