missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ MabSelect™ SuRe™ LX Columns
Polyclonal antibodies, monoclonal antibodies (mAb), and antibody fragments are usually purified by affinity chromatography. Resins containing an immobilized ligand (e.g., protein A, protein G, or protein L) are used to capture antibodies and antibody fragments.
Brand: Cytiva 29157185
This item is not returnable.
View return policy
Description
- Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
- High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
- Generic elution profile for different monoclonal antibodies enables platform approach to purification
- High-flow agarose matrix allows the processing of large feed volumes
- Reduces the risk of product contamination and carryover
- Improves performance and reduces overall costs
Specifications
Antibody Purification Column | |
20 mL/min. (Max) | |
85 μm | |
5 mL | |
5 to 150 mg/Column | |
5/Pk. |
MabSelect SuRe LX | |
Protein A | |
Human IgG | |
Purification of Human IgG | |
2°C to 8°C |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction