missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ MabSelect™ SuRe™ LX Columns
Polyclonal antibodies, monoclonal antibodies (mAb), and antibody fragments are usually purified by affinity chromatography. Resins containing an immobilized ligand (e.g., protein A, protein G, or protein L) are used to capture antibodies and antibody fragments.
£840.00 - £4318.00
Specifications
Product Type | Antibody Purification Column |
---|---|
Stationary Phase | MabSelect SuRe LX |
Ligand | Protein A |
Particle Size | 85 μm |
Purification Target | Human IgG |
Product Code | Brand | Volume (Metric) Bed | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Bed | Quantity | Price | Quantity & Availability | ||||
16437695
|
Cytiva
29268402 |
1 mL | 5/Pk. |
£840.00
Pack of 5 |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16417695
|
Cytiva
29130609 |
1 mL | 10/Pk. |
£2723.00
Pack of 10 |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16427695
|
Cytiva
29157185 |
5 mL | 5/Pk. |
£4318.00
Pack of 5 |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
- High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
- Generic elution profile for different monoclonal antibodies enables platform approach to purification
- High-flow agarose matrix allows the processing of large feed volumes
- Reduces the risk of product contamination and carryover
- Improves performance and reduces overall costs
Specifications
Antibody Purification Column | |
Protein A | |
Human IgG |
MabSelect SuRe LX | |
85 μm | |
Purification of Human IgG |