missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ NHS-Activated HP Columns

Product Code. p-3667224 Shop All Cytiva Products
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
Flow Rate:
20 mL/min.
4 mL/min.
Bed Volume:
1mL
5mL
Diameter (Metric) Inner:
0.7cm
1.6cm
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 10320075

Brand: Cytiva 17071601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Complete instructions for use and method optimization, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems

Couple ligands containing primary amines easily. Cytiva Life Sciences™ HiTrap™ NHS-Activated HP Columns are prepacked with NHS activated Sepharose™ High Performance.

Prepacked with NHS-activated Sepharose™ High Performance Media for coupling ligand via primary amines at close to neutral pH

  • Designed for use with a syringe, peristaltic pump, or chromatography system
  • Degree of substitution: 10 μmol NHS/mL medium
  • Delivered in 100% isopropanol to protect the active groups
  • Stable in the pH range 3 to 12, depending upon ligand
  • Average particle size: 34μm

Open in Edit Popup
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 1mL
Flow Rate 4 mL/min.
Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 1mL
Product Type NHS-Activated HP Column
Diameter (Metric) Inner 0.7cm
For Use With (Application) Preparative affinity chromato graphy, easy coupling of ligands containing primary Amines
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type 10μM NHS groups/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 3 to 12
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 8°C
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.