missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein A HP Prepacked Columns

Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. Cytiva Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.

Brand:  Cytiva 17-0402-03

Code : GE

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10146184

  • £315.00 / Pack of 2
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Prepacked with Protein A Sepharose High Performance media for purification and fractionation of IgG subclasses
  • For use with syringe, pump or chromatography system such as KTA design or FPLC System
  • Binds 20mg human IgG/mL medium
  • Stable over pH 3 to 9 for use and 2 to 10 for cleaning
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • 1 and 5mL columns

Media
  • Ligand: Native protein A
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

20 mg/mL IgG Medium
Chromatography System, Manual
4°C to 8°C, 20% Ethanol
<4 mL/min.
For syringe, peristaltic pump or a chromatography system
2 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers