missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Protein A HP Prepacked Columns
Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. Cytiva Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.
£315.00 - £2972.00
Specifications
Binding Capacity | 20 mg/mL IgG Medium |
---|---|
For Use With (Application) | Chromatography System, Manual |
For Use With (Equipment) | For syringe, peristaltic pump or a chromatography system |
Content And Storage | 4°C to 8°C, 20% Ethanol |
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
11768438
|
Cytiva
17-0403-03 |
5 x 5 mL | <20 mL/min. |
£2972.00
Pack of 5 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10676315
|
Cytiva
17-0402-01 |
5 x 1 mL | <4 mL/min. |
£698.00
Pack of 5 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
10146184
|
Cytiva
17-0402-03 |
2 x 1 mL | <4 mL/min. |
£315.00
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11758438
|
Cytiva
17-0403-01 |
1 x 5 mL | <20 mL/min. |
£663.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
Description
- Prepacked with Protein A Sepharose High Performance media for purification and fractionation of IgG subclasses
- For use with syringe, pump or chromatography system such as KTA design or FPLC System
- Binds 20mg human IgG/mL medium
- Stable over pH 3 to 9 for use and 2 to 10 for cleaning
- Designed for use with a syringe, a peristaltic pump, or a chromatography system
- Maximum pressure: 3 bar
- Column material: polypropylene
- 1 and 5mL columns
Media
- Ligand: Native protein A
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: 3 to 9
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
20 mg/mL IgG Medium | |
Chromatography System, Manual | |
For syringe, peristaltic pump or a chromatography system | |
4°C to 8°C, 20% Ethanol |