missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein G HP Prepacked Columns

Use for fast, easy separations of IgG from different species and subclasses. Cytiva Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.

Brand:  Cytiva 17-0405-03

Code : GE

Additional Details : Weight : 0.12330kg

 View more versions of this product

Product Code. 11584895

  • £3238.00 / Pack of 5
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Prepacked with Protein G Sepharose High Performance
  • Binding capacity: 25mg human IgG/column
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • For use with syringe, peristaltic pump, or chromatography system
  • 1 and 5mL columns

Media
  • Ligand: Recombinant protein G lacking albumin-binding region
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

Pre-packed Columns
16 mm
>20 mL/min.
Designed for use with a syringe, peristaltic pump, or chromatography system
20% Ethanol
5 Pk.
125 mg Human IgG/Column
0.62 in.
5 mL
HiTrap Protein G HP are prepacked columns for general-purpose preparative purification of monoclonal and polyclonal antibodies from most species including rat.
4°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers