Learn More
Cytiva HiTrap™ Q Sepharose Fast Flow IEX Columns
Use for small-scale protein purifications. Cytiva Life Sciences™ HiTrap™ Q Sepharose Fast Flow IEX Columns are strong anion exchangers for rapid separations either alone or connected in series.
Brand:  Cytiva 17-5053-01
 View more versions of this product
Product Code. 10607275
  • £136.00 / Pack of 5
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap Q FF
<4 mL/min.
4°C to 30°C in 20% Ethanol
1 mL
0.2 in.
For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules
7 mm