missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ SP Sepharose Fast Flow IEX Columns
Use for small-scale protein purifications Cytiva Life Sciences™ HiTrap™ SP Sepharose Fast Flow IEX Columns are strong cation exchanger columns for rapid separations either alone or connected in series.
Brand: Cytiva 17-5054-01
This item is not returnable.
View return policy
Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic system
- High flowrates and good scale-up potential
Specifications
HiTrap SP FF | |
<4 mL/min. | |
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol | |
1 mL |
0.2 in. | |
For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules | |
7 mm | |
5 x 1 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap SP FF 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction