missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HM 650V Vibrating Blade Microtome Accessories

Brand:  Epredia™ 350220

Code : NEW

Additional Details : Weight : 0.99000kg

 View more versions of this product

Product Code. 10434150

  • £49.00 / Each
Estimated Shipment: 23-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier
  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
StemiScope™ with optics and mount
  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide
  • 3.5mm fiber bundle
  • Swan-neck light guide: 50mm long
  • Flexible light guide: 750mm long
  • High light intensity
  • Include adapter for the KL200 (760317)

Specifications

Specifications

Cyanacrylat Glue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers