missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HM 650V Vibrating Blade Microtome Accessories
£49.00 - £271.00
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12883123
|
Epredia™
419150 |
Microtome Blade Holder |
£271.00
Each |
Estimated Shipment: 04-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434150
|
Epredia™
350220 |
Cyanacrylat Glue |
£49.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12093039
|
Epredia™
350220 |
Cyanacrylat Glue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Standard buffer tray
- Holds stages in place using a magnet
- Fill tray with all water-based buffers and solutions
- Use pre-cooled in the standard buffer tray for cooling tissue
- Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
- Magnification 2.5X
- Move, swivel, and displace for optimal viewing position
- Includes adapter and illumination
- Eyepieces: 10X
- Supplementary lens: 0.63X
- Zoom magnifying range: 4.1 to 31.5X
- Microscope adapter for angled view of specimen
- Horizontal view and vertical drive and swivel mechanism
- 3.5mm fiber bundle
- Swan-neck light guide: 50mm long
- Flexible light guide: 750mm long
- High light intensity
- Include adapter for the KL200 (760317)