missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units
For use with Mighty Small electrophoresis units
Brand: Hoefer™ SE255D
Code : NEW
Additional Details : Weight : 0.09000kg
Description
- Designed for quick screening and rapid separation
Specifications
SE 260 | |
Buffer Chamber |