missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units
For use with Mighty Small electrophoresis units
£107.00 - £632.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10300455
|
Hoefer™
SE255D |
SE 260 | N/A | |||||||
10561443
|
Hoefer™
SE255D |
SE 260 |
£125.00
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||||
12084757
|
Hoefer™
SE254B |
SE 250 or SE 260 |
£447.00
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||||
10350065
|
Hoefer™
SE254B |
SE 250 or SE 260 | N/A | |||||||
12024877
|
Hoefer™
SE255 |
SE 250 |
£107.00
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||||
Description
- Designed for quick screening and rapid separation
Specifications
Buffer Chamber |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units