missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Brand:  Hoefer™ SE254B

 View more versions of this product

Product Code. 12084757

  • £447.00 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed for quick screening and rapid separation
Specifications

Specifications

SE 250 or SE 260
Buffer Chamber
Foam gasket
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers