Learn More
Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Rubber Overlay Sheet
Dries acrylamide and agarose gels rapidly, evenly and safely.
Brand:  Hoefer™ SE1143
Product Code. 10848910
Description
Description
  • Clear Silicone Rubber Overlay Sheet
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Rubber Overlay Sheet
Silicone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Rubber Overlay Sheet > Overlay sheet