missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Clavies™ Biohazard Bag Holders

Keep bags open and ready to use

Brand:  Bel-Art™ F13192-0001

 View more versions of this product

Product Code. 11322154

  • £97.35 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Economical, lightweight stainless-steel wire frame
  • Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
  • Tray serves as containment well in case of bag leakage
  • Autoclavable at 121°C (250°F) and dishwasher safe
  • Assembly is easy and permanent
  • Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications

Specifications

Biohazard Bag Holder
Silver, Orange
Designed to securely support the bag and its contents
Stainless steel wire frame and orange polypropylene bottom, plastic wire anchors
Sterile
Small
61 cm
47 cm
Autoclavable
Reusable
01-826A, 01-826-5, 01-827B, 01-814-1, 01-815A small 30 x 61cm Clavies™ biohazard bag
Stainless Steel
Benchtop
24 in.
18.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Clavies™ Biohazard Bag Holders > Size: Small

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.