missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Mini Pictograms Hazardous Labels
Brand: Brady™ 814022
14.23 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Gloss | |
96/Pk. | |
Pipe markers and safety signs | |
0.061 mm | |
-18°C to 60°C | |
Toxic Material | |
Black on White |
MINI PIC1809-10X10-GHS06/96 | |
Vinyl | |
Permanent Acrylic | |
10 mm | |
105 mm | |
Diamond | |
Evacuation Panel Safety Sign |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
27Hours
17Minutes
35Seconds
26
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Vinyl Mini Pictograms Hazardous Labels > Toxic Material
Spot an opportunity for improvement?Share a Content Correction