missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Glass Plates for Vertical Electrophoresis Units
Glass plates for vertical electrophoresis units
Brand: Hoefer™ SE6402
This item is not returnable.
View return policy
Description
Glass plates for vertical electrophoresis units
- For use with Hoefer SE600 and SE400 Series Vertical Electrophoresis Units
- Storage temperature: +15°C to +30°C
Specifications
SE 600, SE 400 Series Vertical Electrophoresis System | |
Glass | |
3.14 in. |
Glass Plate | |
18 x 8cm |