missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Glass Plates for Vertical Electrophoresis Units

Glass plates for vertical electrophoresis units

Brand:  Hoefer™ SE6602

 View more versions of this product

Product Code. 12014867

  • £129.29 / Pack of 2
Estimated Shipment: 21-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Glass plates for vertical electrophoresis units

  • For use with Hoefer SE600 and SE400 Series Vertical Electrophoresis Units
  • Storage temperature: +15°C to +30°C
Specifications

Specifications

SE 600, SE 400 Series Vertical Electrophoresis System
Glass
9.4 in.
Glass Plate
18 x 24cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers