missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems

Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

Brand:  Hoefer™ SE260-10A-1.5

 View more versions of this product

Product Code. 15182345

  • £721.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: SE250 Basic System includes: upper buffer chamber, lower buffer chamber, interlocking safety lid with leads, ten 10 x 8cm glass plates, two 10 x 8cm notched alumina plates, four spring clamps, gel seal, well locating decal, and Spacer Mate. SE250 Complete System also includes: Dual Gel Caster, two 0.75mm thick 10-well combs, and two pairs 0.75mm thick spacers. SE260 Basic System includes same items as SE250 Basic Systems, except the glass and alumina plates.SE260 Complete Systems add Dual Gel Caster, ten 10.5 x 10cm glass plates, five 10.5 x 10cm notched alumina plates, and two 10-well combs and two pairs spacers (either 0.75mm or 1.5mm thick; see ordering table).
This item is not returnable. View return policy

Description

Description

Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

  • Run two gels at once
  • Produce high-resolution separations in as little as 45 minutes with no band “smiling”
  • Use alumina plates (included with specified models) for efficient cooling
  • Buffer volume: 300 mL

Applications:

Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis

Certifications

CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.

Specifications

Specifications

EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.15 cm), well locating decals, gel seal
45°C
0.05 in.
10.14 oz.
3.14/4.13 in.
6.49 in.
16.5 cm
7.08 in.
6.29 in.
500 V
500 mA
Up to 80%
113°F
15°C to 30°C
0.15 cm
300 mL
8/10.5 cm
3.93 in.
10 cm
18 cm
16 cm
12 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers