missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units

Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units

Brand:  Hoefer™ SE6119-2-.75

 View more versions of this product

Product Code. 12024717

  • £65.70 / Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ranges from 0.75 to 3.0 mm Thickness
  • 8, 16 and 24 cm in Length
  • 1 to 2 cm Width
  • 2 Pk.
Spécification

Spécification

SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
2 x 16 cm x 0.75mm
0.02 in.
16 cm
Spacer
2/Pk.
0.07 cm
2 cm
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Product Certifications
Promotions

Promotions