missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Holder Clamp

OHAUS™ Holder Clamp provides secure support for holding glassware at elevated positions.

Brand:  OHAUS™ 30392311

 View more versions of this product

Product Code. 15752359

  • £229.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel is autoclavable and corrosion-resistant
  • Aluminum is lightweight yet strong
  • Cast iron with black enamel finish is chemical-resistant
  • Available in different shapes and sizes to fit your laboratory needs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fixed 90° Connection Angle
Aluminum
7.75 in.
Holder
Clamp, Holder, Shelf Grip, CLC-SHGRPA
197.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.